Learn More
BioworldSupplier Diversity Partner NeuroPlex LO Serum-Free Neuronal Medium, 500 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306111781
Like Neurobasal™, Gemini's NeuroPlex ™LO serum-free neuronal medium is intended for the long-term cultureof embryonic hippocampalneurons; NeuroPlex ™ LO also performs well with many other cells from the central and peripheral nervous systems. This specialized medium is intended for supplementation withGem21 NeuroPlex ™, G5 Neuroplex ™, or N2 NeuroPlex ™, depending upon targeted cell type..Sterile-filtered Ships on ice. Storerefrigerated at 2 to 8°CUsage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.
Catalog No. 50-254-248