Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurturin Mouse anti-Human, Alexa Fluor™ 532, Clone: 32000000000000000000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222024AF532

Catalog No. NP222024A32


Add to cart

Description

Description

Neurturin Monoclonal antibody specifically detects Neurturin in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Neurturin
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
neurturin, NTN
Mouse
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Western Blot, ELISA
Conjugated, Purified
NRTN
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant human Neurturin. Mouse anti human Neurturin Q99748.
Protein G purified
Cytokine Research, Immunology
Primary
4902.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit