Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurturin Mouse anti-Human, Alexa Fluor™ 594, Clone: 32000000000000000000000, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222024AF594
Description
Neurturin Monoclonal specifically detects Neurturin in Human samples. It is validated for Western Blot, ELISA.Specifications
Neurturin | |
Monoclonal | |
Western Blot, ELISA | |
neurturin, NTN | |
Mouse | |
Protein G purified | |
Cytokine Research, Immunology | |
4902.0 | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
NRTN | |
Human recombinant Neurturin | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title