Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutec™ Masticator Paddle Lab Blenders

Used for solid and semisolid samples by applying pressure to samples within sterile, disposable bags

Manufacturer:  Neutec GroupSupplier Diversity Partner 9000461

 View more versions of this product

Catalog No. 14-910-428


  Check Availability
Add to cart

Specifications

Description

Warranty

3 years

Description & Specifications

Specifications

Weight: 20kg (44 lb.)
500 - 1500rpm
Digital
39.8 x 23.8 x 26cm (15.7 x 9.4 x 10.2 in.)
Lab Blender
Silver
5/6/7/8
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit