Learn More
Getinge USA Instrument Lubricant Plus Liquid 1 gal. Jug Mild Scent, 4/case
Manufacturer:  Getinge USA 61301605247
Encompass_Preferred
Extends the life of an instrument and ensures proper working of hinges, pins, joints and box locks. Steam and eto permeable. Easy to use; no rinsing or wiping required. Applies a thin film of lubricant to prevent rusting, discoloration and corrosion
Catalog No. NC9152350