Learn More
Human/Mouse AP3B2 (aa 260-274), Synthetic Peptide, Invitrogen™
Synthetic Peptide
Manufacturer:  Invitrogen PEP294
Catalog No. 01-670-214
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Human and Mouse Beta-NAP (residues 260-274) Synthetic Peptide, PEP-294, from Invitrogen.
PEP-294 is a mixture of two, 15 amino acid synthetic peptides whose sequences correspond to residues 260-274 of human and mouse beta-NAP. The sequence of these peptides is (amino to carboxyl terminus): L (260) L E E N A/P E K A F Y G S E E (274).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, beta-NAP, catalog # PA1-1067. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (~70-fold) for competitive inhibition of antibody-protein binding reactions.
Specifications
Specifications
Human, Mouse
11775, 8120
Human, Mouse
Control
peptide with no preservative
L(260) L E E N A/P E K A F Y G S E E(274)
-20° C, Avoid Freeze/Thaw Cycles
Adapter-related protein complex 3 beta-2 subunit; Beta3B-adaptin; Adaptor protein complex AP-3 beta-2 subunit; AP-3 complex beta-2 subunit; Clathrin a
AP3B2
Control
Synthetic Peptide
HPLC
50μg
Liquid
N
RUO
No preservative
Human/Mouse AP3B2 (aa 260-274)
Q13367, Q9JME5
Peptide