Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neutrophil Elastase/ELA2 Mouse anti-Human, Clone: AHN-10, Novus Biologicals™
Mouse Monoclonal Antibody
$212.24 - $608.00
Specifications
Antigen | Neutrophil Elastase/ELA2 |
---|---|
Clone | AHN-10 |
Dilution | Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Radioimmunoassay 1:100-1:2000 |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays |
Classification | Monoclonal |
Description
Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Radioimmunoassay.Specifications
Neutrophil Elastase/ELA2 | |
Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Radioimmunoassay 1:100-1:2000 | |
Monoclonal | |
Ascites | |
RUO | |
Human | |
P08246 | |
1991 | |
This antibody is specific to Neutrophil Elastase | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
AHN-10 | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays | |
Unconjugated | |
Mouse | |
Cancer, Neuroscience | |
Ascites with No Preservative | |
Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
ELANE | |
IgG1 | |
Unpurified | |
Reacts specifically with human neutrophil elastase and not with the 3 homologous neutrophil neutral serine proteases, cathepsin G, esterase N and proteinase 3. Inhibits the elastinolytic activity of elastase. Elastase is felt to play an important role in a number of disease states. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title