Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutrophil Elastase/ELA2 Mouse anti-Human, PE/Atto594, Clone: 950334, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals MAB9167PEATT594

Catalog No. M9167PET594

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Neutrophil Elastase/ELA2 Monoclonal antibody specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Neutrophil Elastase/ELA2
950334
Flow Cytometry
Conjugated, Purified
ELANE
Partial recombinant human ELA2 protein (aa28-252)
Protein A or G purified
Cancer, Neuroscience
Primary
1991.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit