Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutrophils Rabbit anti-Human, Mouse, Rat, Clone: FUT4/1478R- Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody

$458.85

Specifications

Antigen Neutrophils
Clone FUT4/1478R
Host Species Rabbit
Isotype IgG Kappa
Target Species Human, Mouse, Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254374100 View Documents Novus Biologicals
NBP254374100UG
100 ug Each for $458.85
Add to cart
 
NB254374200 View Documents Novus Biologicals
NBP254374200UG
200 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

SSEA-1 Monoclonal antibody specifically detects SSEA-1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications

Specifications

Neutrophils
Rabbit
Human, Mouse, Rat
RUO
Purified
Primary
PBS with No Preservative
FUT4/1478R
IgG Kappa
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Store at -20 to -80C. Avoid freeze-thaw cycles.
Monoclonal
1 mg/mL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit