Learn More
Neutrophils Rat anti-Mouse, Alexa Fluor 405, Clone: RM0028-3G23, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212439AF405
Catalog No. NB212439405
Add to cart
Description
Description
Neutrophils Monoclonal specifically detects Neutrophils in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Neutrophils
RM0028-3G23
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
Isolated mouse neutrophil cells
Protein G purified
Store at 4C in the dark.
Monoclonal
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Rat
IgG2
100 ul
Primary
Mouse