Learn More
Neutrophils Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0028-3G23, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212439AF532
Catalog No. NP212439A32
Add to cart
Description
Description
Neutrophils Monoclonal antibody specifically detects Neutrophils in Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
Neutrophils
RM0028-3G23
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Isolated mouse neutrophil cells
0.1 mL
Primary
Mouse