Eppendorf™ Flask Platforms for Model C76 Water Bath Shaker
Dedicated-use platforms for one specific flask size
Manufacturer: Eppendorf™ M1231-9934
 View all options for this product
Catalog No. 14-284-34
  Check Availability
Add to Cart
Required Accessories : Use of half-size platforms requires subplatform (14-284-46); order separately.
    Capacity (Metric) 125mL
    For Use With (Equipment) Model C76 water bath shaker
    Includes 22 Clamps
    Dimension (Metric) Exterior 42 x 31cm
    Product Type Erlenmeyer flask dedicated platform
    Choose one full-size (16.5L x 12 in.W [42 x 31cm]) platform or two half-size platforms for shaking versatility.
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback