Learn More
Eppendorf™ Gable Covers for Model C76 Water Bath Shaker Available on GSA/VA Contract for Federal Government customers only.
For C76 Water Bath Shaker
Manufacturer:  Eppendorf™ M12312000
 View more versions of this product
Catalog No. 14-284-36
Add to cart
Description
Description
  • Plexiglas™ cover permits temperatures up to 60°C Stainless-steel cover required for temperatures above 60°C
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Model C76 water bath shaker
Stainless Steel