Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ New Brunswick™ Innova™ 42/42R Benchtop Stackable Incubator Shaker

For total versatility, the New Brunswick™ Innova™ 42/42R (refrigerated) Benchtop Stackable Incubator Shaker can be placed in numerous locations, and has user options including a static culture drawer, gassing manifold, remote alarm, and humidity monitor and drip pan.

Supplier:  Eppendorf™ M13350002

 View more versions of this product

Catalog No. 05-400-156


Add to Cart

Alert:

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.


Description

Description

  • High capacity in a small footprint: For example, 4 x 5 L or 2 x 6 L flasks on universal platform, device footprint 63.5 cm (25 in.) width, 74.5 cm (9.3 in.) depth
  • Speed Range: 25 to 400 rpm, 25 to 300 rpm when using flasks, >500 mL in stacked devices
  • Ergonomical and safe interface handling: Top mounted controls and display make bending down unnecessary during handling, avoids accidental parameter changes, and can be reached easily even in a double stack
  • Easy to clean spill diverter and drainable built-in spill pan with rounded corners protect the drive from accidental spills (can be used as a water reservoir to reduce sample evaporation), optional airflow UV-germicidal light
  • Lab position flexibility: Lab position on or under bench, 2x stackable
  • Parallel static cultivation: One height adjustable static shelf included, optional culture drawer and additional shelves available
  • Sample evaporation protection: Bottom spill pan usable as water reservoir, optional culture drawer, for Agar plates, for example
  • Visual sample control: Large viewing window and internal light provide easy viewing of samples without the need to open the door
  • Reliable 24/7 with high loads: Proven and tested Innova™ triple eccentric counterbalanced drive (three weight-supporting shafts), max. load 15 kg
  • Programmability: Continuous, timed, or multi-step programming (15 steps), store up to four programs
  • Options and Accessories: Orbit 19 or 25 mm, photosynthetic light bank, UV germicidal light, culture drawer for evaporation protection of Agar plates, additional static shelves (one included), humidity monitoring, remote monitoring with VisioNize™ Lab Suite and more
Specifications

Specifications

1.9 cm (0.75 in.)
25 to 400 rpm
230 V
1500 VA (12.5 A)
20 to 35 lb.
±0.25°C at 37°C
Aluminum Platform
20.3 in.
51.5 cm
46 × 46 cm (18 × 18 in.)
32.2 in.
81.8 cm
25 in.
63.5 cm
217 lb.
±1 rpm
LED Digital Electric Display
20 to 80%, Non-condensing
USB, Ethernet
Universal/Dedicated
3 Year
9 to 15.8 kg
120 V, 60 Hz
Ambient plus 5°C to 80°C
6 L Flasks
2014/35/EU: EN 61010-1, EN 61010-2-010, EN 61010-2-051, 2014/30/EU: EN 61326-1, EN 55011 (Class A), 2011/65/EU: EN IEC 63000, IEC 61010-1, IEC 61010-2-010, IEC 61010-2-051, UL 61010-1, UL 61010-2-010, UL 61010-2-051, CAN/CSA C22.2 No. 61010-1-12, CSA C22.2 No. 61010-2-010, CSA C22.2 No. 61010-2-051, IEC 61326-1, CISPR 11, FCC 47 CFR Part 15 (Class A)
60 Hz
29.3 in.
74.5 cm
51.5 x 51.7 x 47.7 cm (20.3 x 20.4 x 18.8 in.)
74.5 × 63.5 × 81.8 cm (29.3 × 25 × 32.2 in.)
18.8 in.
47.7 cm
20.4 in.
51.7 cm
98.5 kg
New Brunswick™ Innova™ 42
Plasmid DNA production and protein expression, Biofuel research, Grow bacteria, yeast, fungus, photosynthetic organisms
Accessories
Universal/Dedicated
0.1 sec. to 99 hr. 59 min.
No
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit