Learn More
Eppendorf™ New Brunswick™ Innova™ 42/42R Benchtop Stackable Incubator Shaker
For total versatility, the New Brunswick™ Innova™ 42/42R (refrigerated) Benchtop Stackable Incubator Shaker can be placed in numerous locations, and has user options including a static culture drawer, gassing manifold, remote alarm, and humidity monitor and drip pan.
Supplier: Eppendorf™ M13350012
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
- High capacity in a small footprint: For example, 4 x 5 L or 2 x 6 L flasks on universal platform, device footprint 63.5 cm (25 in.) width, 74.5 cm (9.3 in.) depth
- Speed Range: 25 to 400 rpm, 25 to 300 rpm when using flasks, >500 mL in stacked devices
- Ergonomical and safe interface handling: Top mounted controls and display make bending down unnecessary during handling, avoids accidental parameter changes, and can be reached easily even in a double stack
- Easy to clean spill diverter and drainable built-in spill pan with rounded corners protect the drive from accidental spills (can be used as a water reservoir to reduce sample evaporation), optional airflow UV-germicidal light
- Lab position flexibility: Lab position on or under bench, 2x stackable
- Parallel static cultivation: One height adjustable static shelf included, optional culture drawer and additional shelves available
- Sample evaporation protection: Bottom spill pan usable as water reservoir, optional culture drawer, for Agar plates, for example
- Visual sample control: Large viewing window and internal light provide easy viewing of samples without the need to open the door
- Reliable 24/7 with high loads: Proven and tested Innova™ triple eccentric counterbalanced drive (three weight-supporting shafts), max. load 15 kg
- Programmability: Continuous, timed, or multi-step programming (15 steps), store up to four programs
- Options and Accessories: Orbit 19 or 25 mm, photosynthetic light bank, UV germicidal light, culture drawer for evaporation protection of Agar plates, additional static shelves (one included), humidity monitoring, remote monitoring with VisioNize™ Lab Suite and more
Specifications
2.5 cm (1 in.) | |
25 to 400 rpm | |
230 V | |
1500 VA (12.5 A) | |
20 to 35 lb. | |
±0.25°C at 37°C | |
Aluminum Platform | |
20.3 in. | |
51.5 cm | |
46 × 46 cm (18 × 18 in.) | |
32.2 in. | |
81.8 cm | |
25 in. | |
63.5 cm | |
217 lb. | |
±1 rpm | |
LED Digital Electric Display | |
20 to 80%, Non-condensing | |
USB, Ethernet | |
Universal/Dedicated | |
3 Year |
9 to 15.8 kg | |
230 V, 50 Hz | |
Ambient plus 5°C to 80°C | |
6 L Flasks | |
2014/35/EU: EN 61010-1, EN 61010-2-010, EN 61010-2-051, 2014/30/EU: EN 61326-1, EN 55011 (Class A), 2011/65/EU: EN IEC 63000, IEC 61010-1, IEC 61010-2-010, IEC 61010-2-051, UL 61010-1, UL 61010-2-010, UL 61010-2-051, CAN/CSA C22.2 No. 61010-1-12, CSA C22.2 No. 61010-2-010, CSA C22.2 No. 61010-2-051, IEC 61326-1, CISPR 11, FCC 47 CFR Part 15 (Class A) | |
50 Hz | |
29.3 in. | |
74.5 cm | |
51.5 x 51.7 x 47.7 cm (20.3 x 20.4 x 18.8 in.) | |
74.5 × 63.5 × 81.8 cm (29.3 × 25 × 32.2 in.) | |
18.8 in. | |
47.7 cm | |
20.4 in. | |
51.7 cm | |
98.5 kg | |
New Brunswick™ Innova™ 42 | |
Plasmid DNA production and protein expression, Biofuel research, Grow bacteria, yeast, fungus, photosynthetic organisms | |
Accessories | |
Universal/Dedicated | |
0.1 sec. to 99 hr. 59 min. | |
No |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.