Eppendorf™ Test Tube Racks for Model C76 Water Bath Shaker
Manufacturer: Eppendorf™ AG7TT16
 View more versions of this product
Catalog No. 14-284-23
  Check Availability
Add to Cart
    Specifications
    Capacity (Metric) 16mm
    Includes Insert
    Material Stainless Steel
    For Use With (Equipment) C76 water bath shaker
    No. of Positions 29.0
  • Description & Specifications
Related Products
Product Content Feedback