Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum 5-Drum InLine Spill Deck

Youngstown 5-Drum InLine Spill Deck connects several modules together to allow spills more room to flow.

Manufacturer:  Youngstown Barrel & Drum 2333

Catalog No. 19-339-065


Add to cart

Description

Description

  • Bulkhead fittings “lock” connected modules together and allow spills to flow from one module to next
  • Utilizes hidden containment bladder for additional sump capacity to meet EPA and UFC spill containment regulations
  • Narrow 25.87 in. depth allows drums to be stored in single row along facility walls
  • Low profile design reduces safety and handling concerns
  • Dimensions: 61 x 81.28 x 14.6cm (24 x 32 x 5.75 in.)
  • Weight: 17.71kg (7 lb.)

Compliance

SPCC, EPA Container Storage Regulation 40 CFR 264.175 And UFC Spill Containment Regulations

Specifications

Specifications

SPCC, EPA Container Storage Regulation 40 CFR 264.175 And UFC Spill Containment Regulations
1 Ea.
17 lb.
110 gal.
Safety
50
InLine Spill Deck
7.71kg
416.4L
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit