Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Bulletin Board Biology Charts
SCI_ED

Suited for the reinforcement and review of standards-based topics.

Specifications

Certifications/Compliance Academic Standard for California, Florida, National, New York, Texas
Classification Science
For Use With (Application) Perfect for the reinforcement and review of standard based topics, provides write-on/wipe-off activities
Grade 6, 7, 8, High school
View More Specs

Products 3
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S08945
SCI_ED
View Documents Promotion Details
NewPath Learning™
944703
Modern Genetic Bulletin Board Chart This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S08954
SCI_ED
View Documents Promotion Details
NewPath Learning™
944909
The Endocrine System Bulletin Board Chart This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S08960
SCI_ED
View Documents Promotion Details
NewPath Learning™
944631
Plant and Animal Cell Bulletin Board Chart This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Size: 12 x 18 in.
  • Laminated and double-sided
  • Side 1 features a graphic overview of the topic
  • Side 2 provides“write-on/wipe-off”activities
Specifications

Specifications

Academic Standard for California, Florida, National, New York, Texas
Science
Perfect for the reinforcement and review of standard based topics, provides write-on/wipe-off activities
6, 7, 8, High school
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit