Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mathematics Posters
SCI_ED

Provide illustrated overviews of key mathematical concepts.

$21.75

Specifications

Certifications/Compliance Academic Standard for California, Florida, National, New York, Texas
Classification Mathematics
Grade 5, 6, 7, 8, High school
View More Specs

Products 6
Catalog Number Mfr. No. For Use With (Application) Product Type Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Product Type Price Quantity & Availability  
S04691
SCI_ED
View Documents
NewPath Learning
336102
Comprehensive coverage of decimals and fractions, place value, ordering and comparing decimals and adding and subtracting decimals All About Decimals Poster
Each for $21.75
Add to cart
 
S04690
SCI_ED
View Documents
NewPath Learning
336101
Provides graphic overview of equal parts of a whole, equivalent fractions, fractions on a number line, mixed numbers and adding and subtracting fractions All About Fractions Poster
Each for $21.75
Add to cart
 
S04693
SCI_ED
View Documents
NewPath Learning
336104
A detailed look at key customary and metric units to include length, capacity, weight and temperature Customary and Metric Units of Measurement Poster
Each for $21.75
Add to cart
 
S04694
SCI_ED
View Documents
NewPath Learning
336105
Provides an illustrated overview of the key concepts of ratio, proportion and percent as well as the relationship between them Ratio, Proportion and Percent Poster
Each for $21.75
Add to cart
 
S04696
SCI_ED
View Documents
NewPath Learning
336107
Includes topics like what is the Pythagorean theorem? How is it applied to problem-solving? A detailed overview of this key geometric concept and its uses The Pythagorean Theorem Poster
Each for $21.75
Add to cart
 
S04695
SCI_ED
View Documents
NewPath Learning
336106
Provides a graphic representation of the formulas used to determine volume along with illustrated examples of prisms, cylinders, cones, spheres and pyramids Finding Volume Poster
Each for $21.75
Add to cart
 
Description

Description

  • Measure 23 × 35 in. (58.4 × 88.9cm)
  • Laminated for years of use

Order Info

Grades 5+

Specifications

Specifications

Academic Standard for California, Florida, National, New York, Texas
Mathematics
5, 6, 7, 8, High school
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit