Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Our Solar System Flip Chart Set
SCI_ED

Catalog No. s04872
Click to view available options
Grade:
6 to 10
This item is not returnable. View return policy
This item is not returnable. View return policy

Graphically Introduce and Review Key Curriculum Topics!

Introduce and review standards-based Science concepts. The set features 10 double-sided, laminated charts, each 12 in. x 18 in. and spiral-bound on a durable, free-standing easel –

  • Side 1 of each chart features a graphic overview of topic
  • Side 2 serves as write-on/wipe-off activity chart

Also includes Activity Guide featuring copy-masters and exercises.

Provides comprehensive coverage of the following topics –

  • Formation of Our Solar System
  • Geocentric and Heliocentric Systems
  • Parts of Our Solar System
  • The Sun
  • Measuring Distances in Space
  • The Inner Planets
  • The Outer Planets
  • Comets, Asteroids and Meteors
  • Pluto and the Kuiper Belt
  • Vocabulary Review

Specifications

Grade 6 to 10
Product Type Our Solar System Flip Chart Set
For Use With (Application) Grades 6 to 10
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.