Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ What is Weather? Learning Guide Products for Science Education

Use engaging graphics and inquiry-based activities to enhance student comprehension on standards-based concepts on Weather

$12.75

Specifications

For Use With (Application) Enhance student comprehension on standards-based concepts on weather
Subject Earth Science
Certifications/Compliance Academic Standard for California, Florida, National, New York, Texas
Grade Grade 3, 4 and 5
Product Type Learning Guide
View More Specs

Products 1
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
S13534S Products for Science Education
View Documents
NewPath Learning™
143307
Each
Each for $12.75
Add to cart
 
Description

Description

Full-color, 36-page guide features self-directed readings, easy-to-follow illustrated explanations; guiding questions; inquiry-based activities; a lab investigation; key vocabulary review; and assessment review questions along with a post-test ž

Topics covered:
  • Thinking about Weather
  • ž Our Atmosphere
  • ž ž The Water Cycle
  • ž ž What is Weather? ž
  • ž Who Studies Weather? ž
  • ž Sky Condition ž
  • ž Types of Clouds ž
  • ž Precipitation ž
  • ž Humidity ž
  • ž Air Temperature ž
  • ž Air Pressure ž
  • ž Wind Speed and Direction
ž
Specifications

Specifications

Enhance student comprehension on standards-based concepts on weather
Academic Standard for California, Florida, National, New York, Texas
Learning Guide
Earth Science
Grade 3, 4 and 5
36-page workbook
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit