Learn More
Fisherbrand™ Newport Industrial Poly Chair with Round Tube Base - High Bench Height with Adjustable Arms and Stationary Glides in Poly
Fisherbrand™ Newport Polyurethane Chairs with Round Tube Base are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, handle in back rest, arms, fixed foot ring, & stationary glides. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned.
$457.50
Specifications
Adjustable Yes
Material Polyurethane
Footrest No
With Armrest Yes
With Backrest Yes
View More Specs
Products 1
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-400-400
View Documents
Black
Each for $457.50
Add to cart
Description
Description
  • Adjustable from 26-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Built in handle for ease of movement
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Black, powder-coated steel base with fixed foot ring
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
No
Yes
No
Polyurethane
Yes
No