Learn More
Fisherbrand™ Newport Industrial Poly Chair with Round Tube Base - High Bench Height with Seat Tilt, Adjustable Arms, and Casters in Poly
Fisherbrand™ Newport Polyurethane Chairs with Round Tube Base are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, handle in back rest, arms, fixed foot ring, and self-braking casters. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned.
$516.50
Specifications
Adjustable Yes
Material Polyurethane
Footrest No
With Armrest Yes
With Backrest Yes
View More Specs
Products 1
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-400-407
View Documents
Black
Each for $516.50
Add to cart
Description
Description
  • Adjustable from 28-38 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Built in handle for ease of movement
  • 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Black, powder-coated steel base with fixed foot ring
  • Dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
No
Yes
Yes
Polyurethane
Yes
Yes