Learn More
Fisherbrand™ Newport Industrial Polyurethane Chair - Desk Height with Adjustable Arms and Casters in Polyurethane Products for Science Education
Fisherbrand™ Newport Polyurethane Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments, handle in the back rest, adjustable arms, and self-braking casters. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned.
$327.00
Specifications
Adjustable Yes
Material Polyurethane
Footrest No
With Armrest Yes
With Backrest Yes
View More Specs
Products 1
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S37042 Products for Science Education
View Documents
Black
Each for $327.00
Add to cart
Description
Description
  • Adjustable from 17.25-22.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Built in handle for ease of movement
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Black, nylon-reinforced fiberglass base
  • Dual-wheel, self-braking, non-marring casters suitable for hard floor
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
No
Yes
No
Polyurethane
Yes
Yes