Learn More
Fisherbrand™ Newport Industrial Polyurethane Chair - Desk Height with Seat Tilt and Casters in Polyurethane Available on GSA/VA Contract for Federal Government customers only.
Fisherbrand™ Newport Polyurethane Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments, handle in the back rest, and self-braking casters. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned.
$303.50 - $350.50
Specifications
Adjustable Yes
Base Material Nylon Reinforced Fiberglass
Material Polyurethane
Footrest No
With Armrest No
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-634
View Documents
Blue
Each for $303.50
Add to cart
14-400-364 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Black
Each for $350.50
Add to cart
Description
Description
  • Adjustable from 17.25-22.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Built in handle for ease of movement
  • 8-degree forward and 3-degree backward locking seat tilt
  • Black, nylon-reinforced fiberglass base
  • Dual-wheel, self-braking, non-marring casters suitable for hard floor
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
Polyurethane
No
Yes
Desk Height
Nylon Reinforced Fiberglass
No
Yes
Yes