Learn More
Fisherbrand™ Newport Industrial Polyurethane Chair with Round Tube Base - Medium Bench Height with Seat Tilt and Casters in Polyurethane Products for Science Education
Fisherbrand™ Newport Polyurethane Chairs with Round Tube Base are height adjustable for 36 in. work surfaces and feature ergonomic adjustments, handle in back rest, fixed foot ring, and self-braking casters. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned.
$314.00
Specifications
Adjustable Yes
Material Polyurethane
Footrest No
With Armrest No
With Backrest Yes
View More Specs
Products 1
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36325 Products for Science Education
View Documents
Black
Each for $314.00
Add to cart
Description
Description
  • Adjustable from 24-31.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Built in handle for ease of movement
  • 8-degree forward and 3-degree backward locking seat tilt
  • Black, powder-coated steel base with fixed foot ring
  • Dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
No
Yes
Yes
Polyurethane
No
Yes