Learn More
Newtex™ Zetex™ Heat Resistant Mitts
Provide protection in temperatures to 1000°F and to 2000°F
$13.99 - $283.50
Save up to 50%
Specifications
Material Dupont™ Kevlar™ Aramid Fiber, Fiberglass
For Use With (Application) Foundries and casting operations, welding and hot works operations; glass and ceramic plants production, power generation; petrochemical and refining, laboratory and testing facilities
View More Specs
Products 5
Catalog Number Mfr. No. Coating Material Length (English) Length (Metric) Temperature (English) Max. Price Quantity & Availability  
Catalog Number Mfr. No. Coating Material Length (English) Length (Metric) Temperature (English) Max. Price Quantity & Availability  
11-392-10
View Documents
Newtex™
2100030
None 11 in. 27.94 cm 1000°F Pair for $126.10
Case of 24 PR for $2,720.00
Add to cart
11-392-57
View Documents
Newtex™
2100039
Vermiculite 23 in. 58.42 cm 2000°F Pair for $283.50
Case of 8 PR for $3,960.00
Add to cart
11-392-55
View Documents
Newtex™
2100037
Vermiculite 11 in. 27.94 cm 2000°F Pair for $172.10
Case of 12 PR for $3,690.00
Add to cart
11-392-60B
View Documents
Newtex™
2100032
None 23 in. 58.42 cm 1000°F Pair for $45.88
Case of 48 PR for $2,202.16
Add to cart
11-392-60A
View Documents
Newtex™
2100031
None 14 in. 35.56 cm 1000°F Pair for $13.99
Save 50% Reg : $27.98
Case of 12 PR for $167.82
Save 95% Reg : $3,316.45
Add to cart
Description
Description
  • Made from 100% premium Zetex texturized fiberglass fabric
  • Double stitched and reinforced
  • Premium wool liner (11-392-55, 11-392-56, 11-392-57 only)
Specifications
Specifications
Dupont™ Kevlar™ Aramid Fiber, Fiberglass
Foundries and casting operations, welding and hot works operations; glass and ceramic plants production, power generation; petrochemical and refining, laboratory and testing facilities