Learn More
DuPont™ NexGen™ Protective Coveralls: With Collar
Affordable high-barrier protection
Manufacturer:  DuPont™ NG120SWHMD002500
 View more versions of this product
Catalog No. 19-088-2138
Add to cart
This item is not returnable. View return policy
Description
Description
Lightweight coveralls (1.8 oz. per sq. yd.) provide excellent barrier protection against nontoxic particle, liquid and spray hazards. True aerosol- and liquid-proof microporous barrier demonstrates >99% holdout of particles down to 0.35μm.
  • Breathable, cloth-like material treated to provide antistatic properties for use in controlled environments
  • Proprietary bilaminate composite fabric provides superior liquid penetration1
  • Compared favorably to leading fabric in physical property tests covering resistance to ripping and tearing2
  • Low linting for use in critical applications
  • Long torso with polyester front zipper measures 26 in. long (66cm)
  • White
1 Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
2 ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
Specifications
Specifications
Medium
White
Open
Collared
Serged
ASTM D751, D1117, D5034, D257, AATCC 127
33.75 to 37.25 in.
85.72 to 94.61 cm
10 mil
32.25 in.
5.3 to 5.7 ft.
Fabric
Yes
Open
Front zipper
Lay down
Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
26.5 in.
67.31 cm
Automotive refinishing, waste cleanup and remediation
81.91 cm
1.6 to 1.73 m
SDS
Documents
Product Certifications
Accessories