Learn More
Moore Medical NexTemp™ Single-Use Thermometer
Manufacturer:  Moore Medical 62776
Catalog No. 19-815-399
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Read in Farenheit only (96-104.8°)
  • Latex-free
  • Will not “fire” prematurely
Compliance
Meets ASTM accuracy requirements.
Specifications
Specifications
96° to 104.8°F
96°F
Single use thermometer
105°F
Yes
Thermometer