Learn More
Nextteq™ Gastec™ Detector Tubes: Carbon Monoxide
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Manufacturer:  Nextteq™ 1LA
 View more versions of this product
Catalog No. 19-026-239
  Check Availability
Add to cart
Specifications
Description
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Description & Specifications
Specifications
Carbon Monoxide
1 to 6 min.
10
0° to 40°C
8 to 1000
0.5 to 10
32° to 104°F