NF 157, Tocris

Selective P2Y11/P2X1 antagonist

Manufacturer: R&D Systems 2450/50

Quantity 50mg

 View more versions of this product

Catalog No. 24-505-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C49H28F2N6Na6O23S6
    Formula Weight 1437.08
    Synonym 8,8'-[Carbonylbis[imino-3,1-phenylenecarbonylimino(4-fluoro-3,1-phenylene)carbonylimino]]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt
    Chemical Name or Material NF 157

    Specifications

    Formula Weight 1437.08
    Quantity 50mg
    Synonym 8,8'-[Carbonylbis[imino-3,1-phenylenecarbonylimino(4-fluoro-3,1-phenylene)carbonylimino]]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt
    Recommended Storage Desiccate at Room Temperature
    Assay Percent Range >98%
    Molecular Formula C49H28F2N6Na6O23S6
    CAS 104869-26-3
    Chemical Name or Material NF 157

    Desiccate at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit