Learn More
NF-H Mouse anti-Human, Mouse, Rat, Porcine, Chicken, Alexa Fluor™ 532, Clone: NF421, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233102AF532
Catalog No. NP233102A32
Add to cart
Description
Description
NF-H Monoclonal antibody specifically detects NF-H in Human,Mouse,Rat,Porcine,Chicken samples. It is validated for ELISA.
Specifications
Specifications
NF-H
NF421
ELISA
200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
This MAb reacts with a 200kDa protein, identified as heavy sub-unit of neurofilaments (NF-H). Neurofilaments make up the main structural elements of axons and dendrites and are found in neurons, peripheral nerves, and sympathetic ganglion cells. Neurofilaments consist of three major subunits with molecular weights of 68kDa (NF-L), 160kDa (NF-M) and 200kDa (NF-H). Anti-neurofilament stains a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-neurofilament. Neurofilaments are also present in paragangliomas as well as adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilament.
ELISA
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
NEFH
A recominant protein fragment specific to Neurofilament 200 kD was used as the immunogen for the NF421 antibody.
0.1 mL
Primary
4744
Chicken, Human, Mouse, Porcine, Rat