Learn More
NF-kB p50/105 Rabbit anti-Human,Mouse,Rat, KHL, Clone: 1N19, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB208225UL
 View more versions of this product
Catalog No. ZRB208225MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
NF-kB p50/105 Monoclonal antibody specifically detects NF-kB p50/105 in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
NF-kB p50/105
1N19
KHL
Nuclear factor NF-kappa-B p105 subunit;DNA-binding factor KBF1;EBP-1;Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1;p50/p105 NFkB
Rabbit
IgG
Protein A Purified
Inflammation and Immunology
Primary
Human, Mouse, Rat
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Purified
NFKB1
KLH-conjugated linear peptide corresponding to 16 amino acids from the internal region of human p105 subunit of NF-kB (C-terminal half of p50 subunit).
∼50 and 105 kDa observed; 105.37 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal