Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-kB p50/105 Rabbit anti-Human,Mouse,Rat, KHL, Clone: 1N19, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen NF-kB p50/105
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB208225MI
View Documents
MilliporeSigma
ZRB208225UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB20824XMI
View Documents
MilliporeSigma
ZRB20824X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

NF-kB p50/105 Monoclonal antibody specifically detects NF-kB p50/105 in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

NF-kB p50/105
KHL
Rabbit
Nuclear factor NF-kappa-B p105 subunit;DNA-binding factor KBF1;EBP-1;Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1;p50/p105 NFkB
KLH-conjugated linear peptide corresponding to 16 amino acids from the internal region of human p105 subunit of NF-kB (C-terminal half of p50 subunit).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse, Rat
NFKB1
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit