Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFIB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 166711AP

Catalog No. 50-173-1102


Add to cart

Description

Description

NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.
Specifications

Specifications

NFIB
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00712, P97863
CTF, HMGIC/NFIB, NF I/B, NF1 B, NFI B, NFI RED, NFIB, NFIB2, NFIB3, Nuclear factor 1 B type, Nuclear factor 1/B, nuclear factor I/B, TGGCA binding protein
Rabbit
IgG
150 μL
-20°C
Primary
18028, 29227, 4781
Western Blot
Unconjugated
NFIB
Liquid
Nfib
NFIB Fusion Protein Ag10165
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit