Learn More
Aviva Systems Biology Corp NFIC Antibody - middle region (P100759_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100759_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFIC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NFIC. Alternate names: CTF, CTF5, MGC20153, NF-I, NFIApplicationWB. Species reactivity: Dog, Human, Pig
Catalog No. 50-014-62702