Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NFIL3/E4BP4 Alexa Fluor 647-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8570R100UG
This item is not returnable.
View return policy
Description
NFIL3 Monoclonal specifically detects NFIL3 in Human samples. It is validated for Flow Cytometry.Specifications
NFIL3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q16649 | |
NFIL3 | |
E. coli-derived recombinant human NFIL3/E4BP4 Lys140-Gly462 Accession # Q16649 | |
RUO | |
4783 | |
Human | |
IgG1 |
Flow Cytometry | |
714401 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
E4BP4, E4BP4IL3BP1, IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A | |
Mouse | |
100 μg | |
Primary | |
Detects human NFIL3/E4BP4 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction