Learn More
NFIL3 Mouse anti-Human, Alexa Fluor 750, Clone: 714401, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8570S100UG
Catalog No. FAB8570S100
Add to cart
This item is not returnable. View return policy
Description
Description
NFIL3 Monoclonal antibody specifically detects NFIL3 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
NFIL3
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q16649
NFIL3
E. coli-derived recombinant human NFIL3/E4BP4 Lys140-Gly462 Accession # Q16649
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human NFIL3/E4BP4 in direct ELISAs.
Flow Cytometry
714401
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
E4BP4, E4BP4IL3BP1, IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A
Mouse
IgG1
RUO
Primary
4783
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)