Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFkB1/NFkB p105 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305867F
Description
NFkB1/NFkB p105 Polyclonal specifically detects NFkB1/NFkB p105 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
NFkB1/NFkB p105 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
DNA-binding factor KBF1, EBP-1, NF-kappa-B, NF-kB1, NFKB1, NFKB-p105, Nuclear factor NF-kappa-B p105 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1, p105 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NFkB1/NFkB p105. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
4790 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction