Learn More
NFKB2 p52/p100 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 155031AP
Catalog No. 50-173-0274
Add to cart
Description
Description
This antibody can bind both p52 and p100 isoforms of NFKB2.
The NFkB transcription factor was originally identified as a protein complex consisting of a DNA binding subunit and an associated protein. The subunit is functionally related to c-Rel p75 and Rel B p68. The p50 subunit was initially believed to be a functionally unique protein derived from the amino-terminus of a precursor designated p105. A cDNA has been isolated that encodes an alternative DNA binding subunit of NFkB. It is synthesized as a protein that is expressed in a variety of cell types and, like p105, undergoes cleavage to generate its NFkB subunit, in this case a protein designated p52 (previously referred to as p49). In contrast to p50 derived from p105, p52 acts in synergy with p65 to stimulate the HIV enhancer in transiently transfected Jurkat cells.
Specifications
Specifications
NFKB2 p52/p100
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q00653, Q9WTK5
DNA binding factor KBF2, H2TF1, LYT 10, LYT10, NFKB2, NFKB2,p100, NFKB2,p52,p100, Oncogene Lyt 10, p100, p52
Rabbit
IgG
150 μL
-20°C
Primary
18034, 4791
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NFKB2
Liquid
Nfkb2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse