Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFKBIL2 Mouse anti-Human, DyLight 405, Clone: 648, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243746V

Catalog No. NBP243746V


Add to cart

Description

Description

NFKBIL2 Monoclonal antibody specifically detects NFKBIL2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

NFKBIL2
648
Western Blot
Purified
TONSL
Recombinant protein encompassing a sequence within the N-terminus region of human NFKBIL2. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
IkappaBR, I-kappa-B-related protein, IKBRFLJ40087, Inhibitor of kappa B-related protein, NF-kappa-B inhibitor-like protein 2, NFKBIL2tonsoku-like protein, Nuclear factor of kappa light polypeptide gene enhancer in B-cellsinhibitor-like 2ikappaBR, tonsoku-like, DNA repair protein
Mouse
IgG1
0.1 ml
Primary
4796
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit