Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFKBIL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550192

Catalog No. PA550192


Add to cart

Description

Description

The antibody detects endogenous levels of total NFKBIL2 protein.

The protein encoded by this gene is thought to be a negative regulator of NF-kappa-B mediated transcription. The encoded protein may bind NF-kappa-B complexes and trap them in the cytoplasm, preventing them from entering the nucleus and interacting with the DNA. Phosphorylation of this protein targets it for degradation by the ubiquitination pathway, which frees the NF-kappa-B complexes to enter the nucleus.
Specifications

Specifications

NFKBIL2
Polyclonal
Unconjugated
TONSL
Liquid
TONSL
Fusion protein corresponding to residues near the C terminal of human tonsoku-like, DNA repair protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
2.2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q96HA7
IKBR, NFKBIL2
Rabbit
IgG
100 μL
-20°C
Primary
4796
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit