Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFPS, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective, non-transportable inhibitor of GlyT1 which displays no activity at GABA or glutamate receptors (IC50 values are 2.8, 9.8 and 56000 nM for hGlyT1, rGlyT1 and rGlyT2 respectively). Enhances the amplitude of the NMDA component of glutamatergic EPSCs.
R-enantiomer also available.
Specifications
Formula Weight | 393.45 |
Percent Purity | >98% |
Quantity | 10 mg |
Chemical Name or Material | NFPS |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction