Learn More
Invitrogen™ NFU1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596576
Description
Immunogen sequence: MAATARRGWG AAAVAAGLRR RFCHMLKNPY TIKKQPLHQF VQRPLFPLPA AFYHPVRYMF IQTQDTPNPN SLKFIPGKPV LETRTMDFPT PAAAFRSPLA RQLFRIEGVK SVFFGPDFIT VTKENEELDW NLLKPDIYAT IMDFFASGLP LVTEETPSGE AGSEEDDEVV AMIKELLDTR IRPTVQEDGG DVIYKGFEDG IVQLKLQGSC TSCPSSIITL KNGIQNMLQF; Positive Samples: BT-474, SW480, THP-1, U-937, Mouse liver, Mouse heart, Mouse skeletal muscle, Rat liver, Rat heart; Cellular Location: Cytoplasm, Mitochondrion, cytosol.
This gene encodes a protein that is localized to mitochondria and plays a critical role in iron-sulfur cluster biogenesis. The encoded protein assembles and transfers 4Fe-4S clusters to target apoproteins including succinate dehydrogenase and lipoic acid synthase. Mutations in this gene are a cause of multiple mitochondrial dysfunctions syndrome-1, and pseudogenes of this gene are located on the short arms of chromosomes 1 and 3. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
NFU1 | |
Polyclonal | |
Unconjugated | |
NFU1 | |
0610006G17Rik; CGI-33; HIRA-interacting protein 5; HIRIP; Hirip5; histone cell cycle regulation defective interacting protein 5; iron-sulfur cluster scaffold protein; mHIRIP5; MMDS1; Nfu; NFU1; NFU1 iron-sulfur cluster scaffold; NFU1 iron-sulfur cluster scaffold homolog (S. cerevisiae); NFU1 iron-sulfur cluster scaffold homolog, mitochondrial; NifU; NIFUC; NifU-like C-terminal domain containing | |
Rabbit | |
Affinity Chromatography | |
RUO | |
27247, 297416, 56748 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9QZ23, Q9UMS0 | |
NFU1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human NFU1 (NP_0010027551). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.