Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFXL1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303196A

Description
The recommended shelf life for this product is 1 year from date of receipt.
NFXL1 gene ontology annotations related to this gene include DNA-binding transcription factor activity; zinc ion binding.Specifications
NFXL1 | |
Polyclonal | |
Unconjugated | |
NFXL1 | |
CDZFP, cytoplasm-distribution zinc finger protein, HOZFP, nuclear transcription factor, X-box binding-like 1, ovarian zinc finger protein, OZFP, up-regulated in colon cancer 5, URCC5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
152518 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q6ZNB6 | |
NFXL1 | |
Between 100 and 150 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title