Learn More
Aviva Systems Biology Corp NFYC Antibody - C-terminal region (P100681_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100681_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFYC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NFYC. Alternate names: HSM, CBFC, HAP5, CBF-C, NF-YC, H1TF2AApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62680