Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NG/Neurogranin Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11435R

Description
Neurogranin is the human homolog of the neuron-specific rat RC3/neurogranin gene. This gene encodes a postsynaptic protein kinase substrate that binds calmodulin in the absence of calcium. The NRGN gene contains four exons and three introns. The exons 1 and 2 encode the protein and exons 3 and 4 contain untranslated sequences. It is suggested that the NRGN is a direct target for thyroid hormone in human brain, and that control of expression of this gene could underlay many of the consequences of hypothyroidism on mental states during development as well as in adult subjects.Specifications
NG/Neurogranin | |
Polyclonal | |
Unconjugated | |
NRGN | |
0710001B06Rik; AI838505; B-50 immunoreactive C-kinase substrate; BICKS; BOS_24593; calmodulin-binding protein; hng; LOC100348879; NEUG(55-78); Neurogranin; neurogranin (protein kinase C substrate, RC3); neurogranin-like; NG; NG/RC3; NRGN; p17; postsynaptic spine 1; protein kinase C substrate; Protein kinase C substrate 7.5 kDa protein; protein kinase C substrate, RC3; Pss1; R75334; RC3 | |
Rabbit | |
Protein A | |
RUO | |
4900 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q92686 | |
NRGN | |
KLH conjugated synthetic peptide derived from human NG/Neurogranin. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction