Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NG2 Rabbit anti-Mouse,Rat, Unconjugated, Clone: 1L2, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen NG2
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB532025MI
View Documents
MilliporeSigma
ZRB532025UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB53204XMI
View Documents
MilliporeSigma
ZRB53204X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

NG2 Monoclonal antibody specifically detects NG2 in Mouse,Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Western Blotting.
Specifications

Specifications

NG2
Unconjugated
Rabbit
Chondroitin sulfate proteoglycan 4;Chondroitin sulfate proteoglycan NG2;HSN tumor-specific antigen
Recombinant extracellular domain of rat Chondroitin sulfate proteoglycan 4 (NG2).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunofluorescence, Western Blot
Purified
Mouse, Rat
Cspg4, Ng2
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit