Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NG2 Rat anti-Mouse, Unconjugated, Clone: 1E6, MilliporeSigma™

Rat Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen NG2
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS19491MI
View Documents
MilliporeSigma
MABS1949100UG
100 μg
Each for $372.00
Add to cart
 
MABS19492MI
View Documents
MilliporeSigma
MABS194925UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

NG2 Monoclonal antibody specifically detects NG2 in Mouse samples. It is validated for ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

NG2
Unconjugated
Rat
Chondroitin sulfate proteoglycan 4;Chondroitin sulfate proteoglycan NG2;Proteoglycan AN2
Oli-Neu cells (murine oligodendroglial precursor cells immortalized by an activated neu tyrosine kinase).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Purified
Mouse
Cspg4, An2, Kiaa4232, Ng2
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit