Learn More
Enzo Life SciencesSupplier Diversity Partner NGAL (mouse) monoclonal antibody (29) (biotin) (50 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner BPDABS04329B005
Clone: 29. Host: Rat. Isotype: IgG1κ. Immunogen: Recombinant mouse NGAL. UniProt: P11672. Species Reactivity: Mouse. Applications: ELISA. Purity: Protein A/G-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.14mM sodium chloride and 15mM sodium azide.
Catalog No. 50-658-932